BR73719
BRAND® filter flask, glass, with lateral socket
capacity 500 mL
Synonym(s):
Bottles, Filter flask, Glass
Select a Size
CHF 56.90
Estimated to ship on30 March 2025
Select a Size
About This Item
CHF 56.90
Estimated to ship on30 March 2025
Recommended Products
material
borosilicate glass
flat bottom flask
Joint
joint ST/NS neck33.7 mm
packaging
pkg of 1 ea
manufacturer/tradename
BRAND 73719
bottle capacity
500 mL
capacity
500 mL
funnel capacity
500 mL
neck diam.
33.7 mm
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service