Documents
BR719805
BRAND® counting chamber BLAUBRAND® Fuchs-Rosenthal
without clips, double ruled
Synonym(s):
BLAUBRAND®, Cell culture, Counting Chambers, Fuchs-Rosenthal
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
material
colorless
glass
sterility
non-sterile
feature
without clips
manufacturer/tradename
BRAND 719805
wells
1
General description
The ruling differs from the customary systems for blood cell counts by its large area of 16 mm2.The ruling shows 16 large squares of 1 mm2. Each large square is subdivided into 16 mini squares with 0.25 mm sides and an area of 0.0625 mm2.This chamber is frequently used for counting cells in the cerebro-spinal fluid.
Features and Benefits
• BLAUBRAND® quality• Outstanding measurement precision• 100% certified quality
Packaging
1 counting chamber, complete with 2 haemacytometer cover glasses, in transparent plastic box.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG