Documents
BR462231
BRAND® rubber sleeve for filter crucibles
O.D. 49 mm
Synonym(s):
Filter, Rubber sleeve
About This Item
UNSPSC Code:
41122800
NACRES:
NB.72
material
EPDM rubber
packaging
pack of 10 ea
manufacturer/tradename
BRAND 462231
O.D.
49 mm
Related Categories
General description
The BRAND® Rubber Sleeve for Filter Crucibles is made from high-quality EPDM rubber, designed to provide a secure and reliable fit for various filter crucibles according to the identification code. This durable sleeve enhances the functionality of filter crucibles by ensuring a tight seal, preventing leaks and contamination during filtration processes.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG