Key Documents
BR28920
BRAND® Aluminum Pipet box
Holds 150-200 mm pipettes
Select a Size
1 EA
CHF 62.90
CHF 62.90
Check Cart for Availability
Select a Size
Change View
1 EA
CHF 62.90
About This Item
UNSPSC Code:
41123300
NACRES:
NB.13
CHF 62.90
Check Cart for Availability
material
aluminum
feature
Holds 150-200 mm pipettes
packaging
pack of 1 ea
manufacturer/tradename
BRAND 28920
General description
Pipette box of aluminum, inner silicone coating for storage and sterilisation of pipettes with different length.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)