BR10014
BRAND® compact burette ′Economy′ SILBERBRAND
capacity 50 mL, with PTFE stopcock and PP tip
Synonym(s):
Burettes, Compact burettes, SILBERBRAND
About This Item
Recommended Products
material
glass
packaging
pack of 2 ea
manufacturer/tradename
BRAND 10014
volume measuring range
50 mL, accuracy: ±0.10 mL
burette L
710 mm
capacity
50 mL
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service