Skip to Content
Merck
All Photos(1)

Documents

BMSMR9600T

Benchmark SmartReader 96 Microplate Absorbance Reader with Incubation

AC/DC input 115 V

Synonym(s):

Microplate reader, Absorbance reader, ELISA, 96 well plate, plate reader

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104405
NACRES:
NB.15

manufacturer/tradename

Benchmark Scientific MR9600-T

parameter

temp. range at Ambient +5 to 50 °C

Looking for similar products? Visit Product Comparison Guide

General description

Intuitive, 7 inch color touch screen control panel

Filter based system with wavelength range from 340 to 750nm

Operates as a stand alone system, with USB flash drive drive for data transfer

Absorbance range: 0.0 to 4.000 Abs

8 channel vertical optical path, with zero dispersion

Integrated, multispeed plate shaker

MR9600T features temperature controlled plate incubation

The SmartReader 96 comes complete with 4 popular filters: 405,450,492,630nm and twenty six additional filters are available separately to cover a full range from 340 to 750nm. The integrated filter wheel allows up to 8 filters to be loaded at once, and filters can be selected during programming

Use the touch screen, or connect a mouse for programming and operation. The menu interface allows the user to choose the wavelength filter, set shaking parameters, enter details of the plate layout, and also choose the details for the reading protocol and data calculations.

Model MR9600T offers precise temperature control for incubation of plates during processing. Temperature can be maintained at 5°C above ambient to 50°C.

Up to two filters can be used for 2 measurements of the sample plate, for calculated comparisons. Kinetic calculations can be performed by setting multiple readings.

The SmartReader 96 is a convenient stand alone reader, all software and the programing system is incorporated into the electronics. However, a convenient USB port on the side allows quick and easy saving of data files to a USB drive, and the universal .csv files can then be shared or transferred to a computer for storage or further processing. A data port can be used for connecting a computer or basic printer.

The plate layout interface allows for quick and intuitive set up of a plate’s well contents. The display shows a graphical representation of the 96 well plate, and the user can label each well as required for: “known standard”, “unknown sample”, “blank well”, “QC control well”, “negative control”, “positive control”, or “empty”. The interface allows highlighting and labeling multiple wells to save time.

Measurement data is available immediately on the screen after a reading, as well as interpretations, calculated analysis and curve fittings.

Application

Reading microplates for ELISA assays

Legal Information

SmartReader is a trademark of Benchmark Scientific, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service