Documents
BAF249760001
Workshields for β-emitters, free standing
W × L × H 20.3 cm (8 in.) × 30.5 cm (12 in.) × 37.5 cm (14.75 in.), size S
About This Item
UNSPSC Code:
26142300
NACRES:
NB.35
size
S
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F24976-0001
W × L × H
20.3 cm (8 in.) × 30.5 cm (12 in.) × 37.5 cm (14.75 in.)
General description
Select shield size to match requirements.
  • 9.5 mm (3/8 in.) thick, glass transparent acrylic shields protect against Beta sources and splashes from lab procedures
  • One piece construction has rounded corner for easy cleaning
  • Non skid feet prevent slipping on benchtop
  • Freestanding design has an angled shield and a flat base with a large work surface