Skip to Content
Merck
All Photos(1)

Documents

BAB605701100

SP Bel-Art® H-B Enviro-Safe® Enviro-Safe Liquid-In-Glass Pocket Thermometer

temp 0-220 °F, metal case (with Window )

Synonym(s):

Enviro-Safe® pocket thermometer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.79

material

metal case (with Window )

feature

graduation 2 °F
immersion level Total Immersion

packaging

pack of 1 ea

manufacturer/tradename

HB Instruments B60570-1100

temp measuring range

0-220 °F
accuracy: ±2 °F

L

145 mm
6.3 in. (160 mm)

Looking for similar products? Visit Product Comparison Guide

General description

EnviroKleen certified thermometers feature green, non-toxic, biodegradable, Enviro-Safe® liquid against lead-free, white back glass.
Total immersion. With green, non-hazardous, liquid fill. 160 mm L with aluminum case. An armor option is available, providing extra shielding for the thermometer.

Features and Benefits

  • EnviroKleen certified thermometers feature green, non-toxic, biodegradable, Enviro-Safe® liquid against lead-free, white back glass.
  • Total immersion pocket thermometers are a compact 160mm (6.3”) long
  • Come with a case to protect from breakage
  • Ideal for field studies, waste water treatment and any “on-the-go” applications
  • Case options include:
  • Plastic Window - encased in plastic with an open window
  • Metal Window - encased in metal with an open window
  • Closed Plastic - housed in removable plastic case
  • Closed Metal - housed in a removable metal case
  • Aluminum Duplex - encased in nickel-plated brass with an open window and housed in a removable aluminum case
  • Manufactured and tested by our USA quality team
  • Traceable to NIST
  • Include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
  • Should you require an individualized Thermometer Calibration Report, custom calibration services are available.

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP
Enviro-Safe is a registered trademark of H-B Instrument Co.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service