Skip to Content
Merck
All Photos(1)

Key Documents

B22704

Sigma-Aldrich

N-Benzylglycine ethyl ester

97%

Synonym(s):

Ethyl (benzylamino)acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NHCH2CO2C2H5
CAS Number:
Molecular Weight:
193.24
Beilstein:
2104816
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

reaction suitability

reaction type: solution phase peptide synthesis

color

colorless to yellow

refractive index

n20/D 1.506 (lit.)

bp

140-142 °C/10 mmHg (lit.)

density

1.031 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CCOC(=O)CNCc1ccccc1

InChI

1S/C11H15NO2/c1-2-14-11(13)9-12-8-10-6-4-3-5-7-10/h3-7,12H,2,8-9H2,1H3

InChI key

ULOLIZHBYWAICY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service