Skip to Content
Merck
All Photos(3)

Key Documents

ALD00594

Sigma-Aldrich

2-Hydroxynicotinaldehyde

97%

Synonym(s):

2-Hydroxy-pyridine-3-carbaldehyde, TDG4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

Assay

97%

form

powder or crystals

reaction suitability

reaction type: C-C Bond Formation
reagent type: catalyst
reagent type: ligand
reaction type: C-H Activation

mp

223 °C

functional group

aldehyde

storage temp.

2-8°C

InChI

1S/C6H5NO2/c8-4-5-2-1-3-7-6(5)9/h1-4H,(H,7,9)

InChI key

DNTYEVWEOFZXFE-UHFFFAOYSA-N

Application

This reagent was used as a catalytic transient directing group by Jin-Quan Yu′s lab for Pd(II)-catalyzed γ-C(sp3)-H arylation to couple free primary amines with aryl iodides. Reactions included Pd(OAc)2 683124 and low catalyst and directing group loading.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Yu program focuses on efficient C–H bond activation for drug synthesis, using simple starting materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service