Skip to Content
Merck
All Photos(1)

Documents

A62205

Sigma-Aldrich

2-Amino-3-methylbenzoic acid

99%

Synonym(s):

3-Methylanthranilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(CH3)CO2H
CAS Number:
Molecular Weight:
151.16
Beilstein:
2359694
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder, crystals or chunks

reaction suitability

reaction type: solution phase peptide synthesis

mp

174-177 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cc1cccc(C(O)=O)c1N

InChI

1S/C8H9NO2/c1-5-3-2-4-6(7(5)9)8(10)11/h2-4H,9H2,1H3,(H,10,11)

InChI key

WNAJXPYVTFYEST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Derek Maclean et al.
Journal of combinatorial chemistry, 6(2), 196-206 (2004-03-09)
The design, synthesis, characterization, and screening of a large, encoded thiazolidinone library are described. Three sets of 35 building blocks were combined by encoded split-pool synthesis to give a library containing more than 42 000 members. Building block selection was
R C Kammerer et al.
Xenobiotica; the fate of foreign compounds in biological systems, 16(7), 681-690 (1986-07-01)
Metabolism of lidocaine in rabbit liver 9000 g supernatant fraction was examined. A capillary g.l.c. assay was developed to separate seven known metabolites of lidocaine, and all seven metabolites were identified in extracts of incubations of lidocaine with rabbit-liver fractions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service