Skip to Content
Merck
All Photos(1)

Key Documents

A5487

Sigma-Aldrich

N-Acetyl-2-chloro-2-deoxyneuraminic acid methyl ester 4,7,8,9-tetraacetate

Synonym(s):

5-(Acetylamino)-2-chloro-2,3,5-trideoxy-D-glycero-β-D-galacto-2-nonulopyranosonic acid methyl ester 4,7,8,9-tetraacetate, N-Acetyl-2-chloro-2-deoxy-β-neuraminic acid methyl ester 4,7,8,9-tetraacetate, Methyl N-acetyl-4,7,8,9-tetra-O-acetyl-2-chloro-2-deoxy-β-neuraminate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
CHF 775.00

CHF 775.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
500 MG
CHF 775.00

About This Item

Empirical Formula (Hill Notation):
C20H28ClNO12
CAS Number:
Molecular Weight:
509.89
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

CHF 775.00


Estimated to ship on01 June 2025


Request a Bulk Order

form

solid

shipped in

dry ice

storage temp.

−20°C

SMILES string

COC(=O)[C@]1(Cl)C[C@H](OC(C)=O)[C@@H](NC(C)=O)[C@@H](O1)[C@H](OC(C)=O)[C@@H](COC(C)=O)OC(C)=O

InChI

1S/C20H28ClNO12/c1-9(23)22-16-14(31-11(3)25)7-20(21,19(28)29-6)34-18(16)17(33-13(5)27)15(32-12(4)26)8-30-10(2)24/h14-18H,7-8H2,1-6H3,(H,22,23)/t14-,15+,16+,17+,18+,20-/m0/s1

InChI key

QGJGGAMXXWIKJR-DCDDNHAVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Versatile sialic acid intermediate for the preparation of neuraminic acid derivatives. N-acetylneuraminic acid is often present as the terminal sugar of glycoproteins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service