Skip to Content
Merck
All Photos(1)

Key Documents

94394

Sigma-Aldrich

Dimethoxy-methyl-octadecylsilane

Wacker Chemie AG, >95.0% n- and iso-isomer basis (GC)

Synonym(s):

Methyl-octadecyl-dimethoxysilane, Octadecyl-methyl-dimethoxysilane, Silane-ODM-dimethoxy

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H46O2Si
CAS Number:
Molecular Weight:
358.67
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

>85.0% n-isomer basis (GC)
>95.0% n- and iso-isomer basis (GC)

form

liquid

manufacturer/tradename

Wacker Chemie AG

density

0.86 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCC[Si](C)(OC)OC

InChI

1S/C21H46O2Si/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-24(4,22-2)23-3/h5-21H2,1-4H3

InChI key

UBCPEZPOCJYHPM-UHFFFAOYSA-N

Other Notes

prices for bulk quantities on request

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

239.0 °F

Flash Point(C)

115 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service