940135
Sodium tetrafluoroborate

electrolyte powder, anhydrous, battery grade, 99% trace metals basis
Synonym(s):
Sodium borofluoride, Sodium fluoroborate
About This Item
grade
battery grade
Quality Level
Assay
99% trace metals basis
form
powder
mp
384 °C (lit.)
solubility
soluble (carbonates: soluble(lit.); glymes: soluble(lit.))
density
2.47 g/mL at 25 °C (lit.)
2.47 g/mL at 25 °C
application(s)
battery manufacturing
SMILES string
[Na+].F[B-](F)(F)F
InChI
1S/BF4.Na/c2-1(3,4)5;/q-1;+1
InChI key
KGJZTOFHXCFQIV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Discover the role of electrolytes in sodium-ion batteries, to enhance performance, safety, and sustainability in energy storage solutions.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service