Skip to Content
Merck
All Photos(1)

Key Documents

939439

Sigma-Aldrich

Hyaluronic acid hydrazide

Synonym(s):

ADH, Adipic acid dihydrazide, HA-ADH, Hyaluronic acid, Hyaluronic acid adipic dihydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C14H23N3O10)n(C14H20N NaO11)m
UNSPSC Code:
12352200

form

powder or chunks (or Fibers)

Quality Level

color

white to faint yellow-brown

storage temp.

2-8°C

SMILES string

OC1[C@H](O[*])[C@@H](C(NN)=O)O[C@H]([C@@H]1O)O[C@H]2[C@H](O)[C@@H](CO)O[C@@H](O[C@@H]([C@@H]3O)[C@@H](C(O[Na])=O)O[C@H]([C@@H]3O)O[C@H]4[C@H](O)[C@@H](CO)O[C@H]([C@@H]4NC(C)=O)[*])[C@@H]2NC(C)=O

General description

Hyaluronic acid hydrazide is a versatile and innovative functionalized natural polymer whereby the carboxylic acid moieties of hyaluronic acid are modified with mono- and polyfunctional hydrazides via controlled synthesis. Hyaluronic acid (HA), linear polysaccharide composed of alternating 1,4-linked units of 1,3-linnked glucuronic acid and N-acetylglucosamine, is a crucial component of the extracellular matrix (ECM). The hydrazide groups are highly reactive with aldehydes, leading to the formation of acyl hydrazone linkages. This is particularly useful for linking glycoproteins or carbohydrates in a site specific manner.

Application

  • Hyaluronic acid hydrazide can be used for a wide range of applications. By harnessing the controlled hydrazide modification of hyaluronic acid, this product can be used to create tailored solutions for various fields, including biomedical research, pharmaceuticals, and tissue engineering.
  • Applications include:
  • Developing new drug delivery systems such as prodrugs
  • Studying cell motility in 3D cell culture scaffolds
  • Engineering biocompatible scaffolds and hydrogels for areas such as cartilage tissue engineering
  • The hyaluronic acid hydrazide can also be used as an intermediate to synthesize other HA derivatives with various functional groups.

Features and Benefits

  • Biocompatible and biodegradable
  • Greater than 60% functionalized
  • Tailorable for tethering additional functional groups or biochemical moieties

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G D Prestwich et al.
Journal of controlled release : official journal of the Controlled Release Society, 53(1-3), 93-103 (1998-09-19)
Controlled modification of the carboxylic acid moieties of hyaluronic acid with mono- and polyfunctional hydrazides leads to biochemical probes, biopolymers with altered physical and chemical properties, tethered drugs for controlled release, and crosslinked hydrogels as biocompatible scaffoldings for tissue engineering.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service