Skip to Content
Merck
All Photos(1)

Documents

937398

Sigma-Aldrich

(tBuXPhos)Pd(p-CH3C6H4(Cl)

new
Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H52ClPPd
CAS Number:
Molecular Weight:
657.64
UNSPSC Code:
12352142
NACRES:
NA.21

form

powder or crystals

Quality Level

reaction suitability

reaction type: Cross Couplings

color

white to faint beige

functional group

(Palladium)
phosphine

storage temp.

room temp

SMILES string

Cl[Pd](C1=CC=C(C)C=C1)[P](C2=CC=CC=C2C3=C(C(C)C)C=C(C(C)C)C=C3C(C)C)(C(C)(C)C)C(C)(C)C

General description

(tBuXPhos)Pd(p-CH3C6H4(Cl) is a G6 Buchwald precatalyst. The G6 precatalysts are air-stable, palladium-based oxidative addition complexes (OACs) that offer an alternative to the previously developed classes of palladacycle precatalysts. Unlike previous generations, the bulkiest biarylphosphine ligand precatalysts are readily prepared. These precatalysts have been shown to be effective for many reactions, including C-N, C-O, and C-F cross couplings.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service