Skip to Content
Merck
All Photos(1)

Documents

929409

Sigma-Aldrich

Vitamin E Succinate-(phenyl-5,7-dimethyl-d6)

≥98 atom % D, ≥95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33D6H48O5
Molecular Weight:
536.82
UNSPSC Code:
12352106
NACRES:
NA.13

isotopic purity

≥98 atom % D

Quality Level

Assay

≥95% (CP)

technique(s)

mass spectrometry (MS): suitable

mass shift

M+6

Packaging

This is a bulk stock product. For informaiton on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service