Skip to Content
Merck
All Photos(1)

Key Documents

92791

Sigma-Aldrich

1-Trimethylsilyl-1,2,4-triazole

purum, ≥97.0% (NT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11N3Si
CAS Number:
Molecular Weight:
141.25
Beilstein:
606382
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥97.0% (NT)

refractive index

n20/D 1.461 (lit.)
n20/D 1.462

bp

74 °C/12 mmHg (lit.)

density

0.989 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)n1cncn1

InChI

1S/C5H11N3Si/c1-9(2,3)8-5-6-4-7-8/h4-5H,1-3H3

InChI key

WPSPBNRWECRRPK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent used for the preparation of phosphorous and phosphoric triazolides; Synthesis of 1,1′-carbonyl-di(triazole)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Dabkowski et al.
Nucleic acids research, 12(23), 9123-9135 (1984-12-11)
The synthesis of a model compound, diphenylphosphoric toluene-p-sulfonic anhydride, an arylsubstituted phosphoric sulfonic mixed anhydride, is described. Using the same procedure a thymidyl substituted derivative was prepared. The phosphoric sulfonic anhydride is the presumed intermediate in oligonucleotide coupling reactions involving
L. Birkofer et al.
Chemische Berichte, 93, 2804-2804 (1960)
H.R. Kricheldorf et al.
Angewandte Chemie (International Edition in English), 88, 337-337 (1976)
J.-L. Fourrey et al.
Tetrahedron Letters, 25, 4511-4511 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service