Skip to Content
Merck
All Photos(1)

Key Documents

92750

Sigma-Aldrich

Dimethyl trimethylsilylmethylphosphonate

≥97.0%

Synonym(s):

[(Trimethylsilyl)methyl]-phosphonic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCH2P(O)(OCH3)2
CAS Number:
Molecular Weight:
196.26
Beilstein:
2412153
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

Assay

≥97.0%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.435 (lit.)
n20/D 1.435

bp

111-114 °C/21.5 mmHg (lit.)

density

1.024 g/mL at 25 °C (lit.)

SMILES string

COP(=O)(C[Si](C)(C)C)OC

InChI

1S/C6H17O3PSi/c1-8-10(7,9-2)6-11(3,4)5/h6H2,1-5H3

InChI key

QBHFBIHXALNDCB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Synthesis of phosphonato-phosphine palladacycles
  • Studying the synthetic applications of phosphoryl-stabilized anions
  • Peterson olefination reactions
  • Dilithiation and transmetalation

Other Notes

Reagent for the preparation of olefinic sugar phosphonates; For the direct methylenation of carbonyl compounds

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A. Sekiguchi et al.
Chemistry Letters (Jpn), 1385-1385 (1978)
H. Paulsen et al.
Chemische Berichte, 108, 1732-1732 (1975)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service