925101
E7820-C3-NH2 hydrochloride
Synonym(s):
N-(3-Aminopropyl)-3-(N-(3-cyano-4-methyl-1H-indol-7-yl)sulfamoyl)benzamide hydrochloride, Crosslinker−E3 Ligase ligand conjugate, DCAF15 protein degrader building block
Select a Size
About This Item
Recommended Products
ligand
E7820
Quality Level
reaction suitability
reactivity: carboxyl reactive
reagent type: ligand-linker conjugate
functional group
amine
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Protein Degrader Building Blocks
Technology Spotlight: DCAF15 Degrader Building Blocks for Targeted Protein Degradation
Other Notes
Legal Information
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service