Skip to Content
Merck
All Photos(5)

Documents

922250

Sigma-Aldrich

Male Luer Plug

single, Fluidic 270, PP

Synonym(s):

Microfluidic accessories

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
42142600
NACRES:
NA.23

Application

The male Luer plugs enable closing of female Luer and Luer lock interfaces on our fluidic platforms. The male Luer plugs allow liquid to be moved with the female Luer interface into the fluidic channels on chip, and the fluidic interface remains closed in order to avoid contamination.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent advances of controlled drug delivery using microfluidic platforms
Li X, et al.
Advanced Drug Delivery Reviews, 128 , 3-28 (2018)
Microfluidic-assisted fabrication of carriers for controlled drug delivery.
Santos H A, et al.
Lab on a chip, 17, 1856-1883 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service