Skip to Content
Merck
All Photos(1)

Key Documents

920975

Sigma-Aldrich

TissueFab® bioink 

(GelAlgHA)ma -UV/365 nm

Synonym(s):

AlgMA, Alginate, Bioink, GelMA, Gelatin, HAMA, Hyaluronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
CHF 523.00

CHF 523.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
CHF 523.00

About This Item

UNSPSC Code:
12352201
NACRES:
NA.25

CHF 523.00


Check Cart for Availability

Request a Bulk Order

Quality Level

description

suitable for 3D bioprinting applications

sterility

sterile-filtered

form

viscous liquid (or gel)

impurities

<5 CFU/g Bioburden (Fungal)
<5 CFU/g Bioburden (Total Aerobic)

color

colorless to pale yellow

particle size

0.2 μm

pH

(6.5-7.5)

application(s)

3D bioprinting

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

TissueFab®- HAMA-AlgMA-UV bioink is an alginate methacrylate based bioink for 3D bioprinting applications. The formulation is optimized for 3D bioprinting of tissues and constructs using extrusion based 3D bioprinters, and can be can be used to bioprint cell-laden hydrogels in desired shape without any supporting material. The crosslinking of printed structures can be done in one step using UV light for further culture and maturation of cells for tissue engineering and regenerative medicine applications.

This bioink is derived from natural polymers − hyaluronic acid, alginate and gelatin. Hyaluronic acid is also known as sodium hyaluronate or hyaluronan, is an anionic, nonsulfated glycosaminoglycan (GAG). Alginate also known as sodium alginate or alginic acid, is a natural biopolymer obtained from brown algae. Alginate is a natural polysaccharide widely used in tissue engineering. Gelatin contains bioactive peptide sequences similar to native extracellular matrix which promotes integrin mediated cell adhesion and MMP sensitive enzymatic degradation which are essential for cellular functions such as migration, proliferation and differentiation.

Features and Benefits

  • Ready-to-use formulation optimized for high printing fidelity and cell viability, eliminating the lengthy bioink formulation development process
  • Step-by-step protocols developed and tested by MilliporeSigma 3D Bioprinting Scientists, no prior 3D bioprinting experience needed
  • Suitable for different extrusion-based 3D bioprinter model

Packaging

10 mL in glass bottle

Legal Information

TISSUEFAB is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Bioinks enable 3D bioprinting of tissue constructs for drug screening and transplantation; select suitable bioinks for specific tissue engineering.

Learn how 3D bioprinting is revolutionizing drug discovery with highly-controllable cell co-culture, printable biomaterials, and its potential to simulate tissues and organs. This review paper also compares 3D bioprinting to other advanced biomimetic techniques such as organoids and organ chips.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service