Skip to Content
Merck
All Photos(2)

Documents

915912

Sigma-Aldrich

Np# HCl

Synonym(s):

1,3-Bis(2,4,7-tribenzhydrylnaphthalen-1-yl)-1H-imidazol-3-ium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C101H77ClN2
Molecular Weight:
1354.16
UNSPSC Code:
12161600
NACRES:
NA.22

form

powder

Quality Level

mp

209-306 °C

Related Categories

Application

Np#HCl is sterically-bulky, easily-accessible NHC ligand that can be used in a broad range of catalytic reactions, including cross-coupling reactions, hydrofunctionalizations and element addition reactions using different metals, including but not limited to Pd, Ni, Au, Ag, Cu, Pt, Ru.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

426.2 - 494.6 °F

Flash Point(C)

219 - 257 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service