Skip to Content
Merck
All Photos(1)

Documents

912778

Sigma-Aldrich

sBOX(iPr)

≥95%

Synonym(s):

Diisopropyl 2,2′-(cyclopropane-1,1-diyl)(4S,4′S)-bis(4,5-dihydrooxazole-4-carboxylate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H24N2O6
CAS Number:
Molecular Weight:
352.38
UNSPSC Code:
12352005
NACRES:
NA.22

Assay

≥95%

form

(Powder or Solid or Chunks)

storage temp.

2-8°C

Related Categories

Application

sBOX(iPr) is a serine-derived chiral bisoxazoline ligand. The additional coordinating functionality provided by the esters enables new reactivity compared to traditional BOX ligands. sBOX(iPr) has been shown to be an optimal ligand for the electrocatalytic cyanophosphorylation as well as hydrocyanation of vinylarenes.

Product can be used with our Synlectro electrochemistry platform (ESYNTH001)

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niankai Fu et al.
Journal of the American Chemical Society, 141(37), 14480-14485 (2019-09-10)
In contrast to the rapid growth of synthetic electrochemistry in recent years, enantioselective catalytic methods powered by electricity remain rare. In this work, we report the development of a highly enantioselective method for the electrochemical cyanophosphinoylation of vinylarenes. A new

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service