Skip to Content
Merck
All Photos(1)

Key Documents

912492

Sigma-Aldrich

Silica dioxide- sorbent nanofiber

Synonym(s):

Nanofibrous inorganic powder structure, SiO2

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CHF 235.00
10 G
CHF 410.00

CHF 235.00


Only 0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
CHF 235.00
10 G
CHF 410.00

About This Item

Linear Formula:
SiO2
CAS Number:
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23

CHF 235.00


Only 0 left in stock

Request a Bulk Order

form

nanofiber

Quality Level

fiber diam.

800-1200 nm , structure: amorphous
Length: continuous fibers

surface area

600-900 m2/g

SMILES string

[Si](=O)=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Application

Silica dioxide- sorbent nanofibers can absorb significantly more water than commercially available silicagel of the same mesoporous character. Applications include adsorbent of water and other polar sorbents, catalyst carrier, filtration, separation, li-ion battery separators, and sensors.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Review discusses recent progress and challenges in ORR catalyst development for PEMFC applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service