Skip to Content
Merck
All Photos(2)

Key Documents

909300

Sigma-Aldrich

Carboxylic acid-poly(ethylene glycol)-b-poly(D,L lactide)

PEG average Mn 5,000, PDLA average Mn 16,000

Synonym(s):

COOH-PEG-PDLA, COOH-PEG-PLA, Carboxylic acid PEG-PDLA, Carboxylic acid-PEG-PLA

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CHF 119.00

CHF 119.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
CHF 119.00

About This Item

Linear Formula:
HO(C3H4O2)m(C2H4O)nC2H3O2
UNSPSC Code:
12352106
NACRES:
NA.23

CHF 119.00


Check Cart for Availability

Request a Bulk Order

form

powder or chunks

mol wt

PDLA average Mn 16,000 (by NMR)
PEG average Mn 5,000 (by NMR)

color

white to tan

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Carboxylic acid-poly(ethylene glycol)-b-poly(D,L lactide) is a functionalized, amphiphilic, diblock copolymer composed of a hydrophilic PEG block and a hydrophobic PLA block. These biodegradable, biocompatible polymers can self-assemble to form nanoparticles, such as micelles and polymersomes, in both aqueous and non-aqueous media. Due to these properties, these polymers are widely used in polymeric nanoparticle formulation to achieve controlled and targeted delivery of therapeutic agents (e.g. APIs, genetic material, peptides, vaccines, and antibiotics). The carboxylic acid functional group on the PEG chain enables rapid and facile surface functionalization, allowing for these materials to be used in applications such as targeted drug delivery.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ren Zhong Xiao et al.
International journal of nanomedicine, 5, 1057-1065 (2010-12-21)
Due to their small particle size and large and modifiable surface, nanoparticles have unique advantages compared with other drug carriers. As a research focus in recent years, polyethylene glycol-polylactic acid (PEG-PLA) block copolymer and its end-group derivative nanoparticles can enhance
R Gref et al.
Science (New York, N.Y.), 263(5153), 1600-1603 (1994-03-18)
Injectable nanoparticulate carriers have important potential applications such as site-specific drug delivery or medical imaging. Conventional carriers, however, cannot generally be used because they are eliminated by the reticulo-endothelial system within seconds or minutes after intravenous injection. To address these

Articles

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service