Skip to Content
Merck
All Photos(1)

Key Documents

905704

Sigma-Aldrich

Dimethylsulfoxonium-(N-Boc(4-piperidinecarbonoyl)methylide

≥95%

Synonym(s):

tert-Butyl 4-(2-(dimethyl(oxo)-sulfaneylidene)acetyl)piperidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H25NO4S
CAS Number:
Molecular Weight:
303.42
UNSPSC Code:
12352005

Assay

≥95%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

131-132 °C

functional group

ketone
sulfoxide

storage temp.

−20°C

SMILES string

O=C(C1CCN(C(OC(C)(C)C)=O)CC1)C=S(C)(C)=O

General description

Sulfoxonium ylides are highly stable carbene precursors, serving as a safer alternative to diazo compounds as they that do not generate gases as byproducts. Sulfoxonium ylides have been shown to be effective in a wide array of transition metal catalyzed C-H functionalization and coupling reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuel Barday et al.
Angewandte Chemie (International ed. in English), 56(42), 13117-13121 (2017-08-31)
The functionalization of carbon-hydrogen bonds in non-nucleophilic substrates using α-carbonyl sulfoxonium ylides has not been so far investigated, despite the potential safety advantages that such reagents would provide over either diazo compounds or their in situ precursors. Described herein are
Gia L Hoang et al.
Tetrahedron, 74(26), 3318-3324 (2018-07-11)
The synthesis of bridgehead N-fused [5,6]-bicyclic heterocycles via rhodium(III)-catalyzed C-H functionalization of C-alkenyl azoles with sulfoxonium ylides is disclosed. Reactions proceeded in good to high yields for a range of aryl, heteroaryl and alkyl sulfoxonium ylides. In addition, 2-alkenyl imidazoles
Kim Søholm Halskov et al.
Organic letters, 20(8), 2464-2467 (2018-03-28)
Azolopyrimidines are efficiently prepared by direct imidoyl C-H bond activation. Annulations of N-azolo imines with sulfoxonium ylides and diazoketones under redox-neutral conditions and alkynes under oxidizing conditions provide products with various arrangements of nitrogen atoms and carbon substituents. We have

Related Content

Ketosulfoxonium ylides serve as versatile carbene equivalents for large-scale reactions, releasing dimethyl sulfoxide biproducts.

Ellman group developed electron-rich phosphine ligands for C-H functionalization and tert-Butanesulfinamide for asymmetric amine synthesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service