Skip to Content
Merck
All Photos(1)

Key Documents

901443

Sigma-Aldrich

Trimethylphosphine solution

0.9 M in 2-methyltetrahydrofuran

Synonym(s):

Trimethylphosphane, PMe3, Trimethylphosphorus

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
CHF 115.00

CHF 115.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
CHF 115.00

About This Item

Linear Formula:
(CH3)3P
CAS Number:
Molecular Weight:
76.08
Beilstein:
969138
MDL number:
UNSPSC Code:
12352128
NACRES:
NA.22

CHF 115.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

reaction suitability

reagent type: ligand
reaction type: Mitsunobu Reaction

reagent type: ligand
reaction type: Wittig Reaction

concentration

0.9 M in 2-methyltetrahydrofuran

density

0.838 g/mL

functional group

phosphine

SMILES string

CP(C)C

InChI

1S/C3H9P/c1-4(2)3/h1-3H3

InChI key

YWWDBCBWQNCYNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Trimethylphosphine is an electron-rich phosphine ligand used in the Mitsunobu reaction. It participates in the transformation of azides into carbamates, aziridines from azidoalcohols, iminophosphoranes and aza-Wittig reaction. This product is offered as a solution in 2-methyltetrahydrofuran for more convenient handling.

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

5.0 °F

Flash Point(C)

-15 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service