901417
N-((1S,2S)-1-((R)-4-Isobutyl-4,5-dihydrooxazol-2-yl)-2-methylbutyl)acetamide
Synonym(s):
Chiral APAO ligand
About This Item
form
powder or crystals
reaction suitability
reagent type: catalyst
reagent type: ligand
reaction type: C-H Activation
storage temp.
−20°C
Application
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Yu program focuses on efficient C–H bond activation for drug synthesis, using simple starting materials.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service