Skip to Content
Merck
All Photos(1)

Key Documents

901047

Sigma-Aldrich

Molybdenum trioxide/PEDOT:PSS ink

viscosity 3 cP 

Synonym(s):

Avantama P-30, Ink for charge selective layers in printed electronics, MoO3/PEDOT:PSS dispersion, MoO3/PEDOT:PSS ink

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

description

Solid content: 0.8 wt% crystalline MoO3/PEDOT:PSS

form

dispersion

color

dark blue

particle size

10-30 nm

viscosity

(25 °C) (6500s-1 shear rate)
3 cP

Application

This ink is for charge selective layers in printed electronics. This ink is for spin-coating, doctor blading, or slot-die.

Caution

  • Storage: In dark at room temperature.
  • Prior to application: Shake well.
  • Drying of deposited films at >100 °C.

Legal Information

Product of Avantama Ltd.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

63.5 °F - closed cup

Flash Point(C)

17.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Advancements in bioelectronics, incorporating self-healing materials for wearable devices, and measuring bioelectric signals to assess physiological parameters.

Professor Rivnay (Northwestern University, USA) discusses using organic mixed conductors as an alternative to efficiently bridge the ionic world of biology with contemporary microelectronics.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service