Skip to Content
Merck
All Photos(1)

Documents

900575

Sigma-Aldrich

3-(1H-Imidazol-1-yl)propionate-1,2,3-13C3 sodium salt

≥99 atom % 13C, ≥97% (CP)

Synonym(s):

3-Imidazol-1-yl propionate-1,2,3-13C3 sodium salt, Imidazole propionate-1,2,3-13C3 sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C3H7N2O2Na
Molecular Weight:
165.10
UNSPSC Code:
12352005

isotopic purity

≥99 atom % 13C

Assay

≥97% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+3

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service