Skip to Content
Merck
All Photos(1)

Key Documents

900149

Sigma-Aldrich

UV curable resin

greener alternative

low viscosity

Synonym(s):

CPS 1030 UV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

Quality Level

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

viscosity

75 cP (Brookfield LDTDV-II digital viscometer with size 31 spindle)

density

1.17 g/mL

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to enabling category of greener alternatives and thus aligns with "Design for energy efficiency". Click here for more information.

Application

  • UV curable very low viscosity resin.
  • Cures rapidly to form a glassy high modulus material.
  • Excellent for use in applications requiring low viscosity, high hardness and low shrinkage stress.
  • Good for some spray coatings requiring no volatile organic compounds (VOCS).
  • Non yellowing.
  • Low oxygen inhibition.
  • Mechanical Properties after curing:
Modulus*: 750 MPa
Strength*: 28 MPa
Elongation*: 6%
Tack Free**: 25 fpm

*Test Resources: tensile tester, 35 mm gauge length, 1.0 mm thickness, 6.5 mm width.
** Cured under stated intensity. Medium pressure mercury bulb with 320-500 nm filter. Adhesion tests were conducted using a cross hatch tape test on films 0.125 mm thick on glass and PET. Material data using ASTM D 638-02a.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the cure time for product 900149?

    1 answer
    1. Product 900149 cures at 10 feet per minute under a 300W H bulb. The cure times would be different with a different light type and power, so customers are advised to provide their specific setup for a more accurate estimate.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service