Skip to Content
Merck
All Photos(1)

Documents

86906

Sigma-Aldrich

Tetrabutylammonium methoxide solution

20% in methanol (NT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(OCH3)
CAS Number:
Molecular Weight:
273.50
Beilstein:
4597539
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

form

solution

concentration

20% in methanol (NT)

impurities

≤0.01% halides (as bromide)

anion traces

sulfate (SO42-): ≤500 mg/kg

storage temp.

2-8°C

SMILES string

C[O-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.CH3O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-2/h5-16H2,1-4H3;1H3/q+1;-1

InChI key

HPVRLMGCBINLBH-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service