Skip to Content
Merck
All Photos(1)

Key Documents

86146

Sigma-Aldrich

Dihexyl sulfosuccinate sodium salt solution

technical, ~80% in H2O

Synonym(s):

Dihexyl sodium sulfosuccinate, Sodium dihexyl sulfosuccinate solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H29NaO7S
CAS Number:
Molecular Weight:
388.45
Beilstein:
3818241
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

description

anionic

grade

technical

mol wt

388.45 g/mol

concentration

~80% in H2O

technique(s)

immunocytochemistry: suitable

impurities

~5% ethanol

density

1.121 g/mL at 20 °C
1.121 g/mL at 20 °C

SMILES string

[Na+].CCCCCCOC(=O)CC(C(=O)OCCCCCC)S([O-])(=O)=O

InChI

1S/C16H30O7S.Na/c1-3-5-7-9-11-22-15(17)13-14(24(19,20)21)16(18)23-12-10-8-6-4-2;/h14H,3-13H2,1-2H3,(H,19,20,21);/q;+1/p-1

InChI key

WVFDILODTFJAPA-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service