Skip to Content
Merck
All Photos(1)

Key Documents

855995

Sigma-Aldrich

6-Bromo-2-naphthyl β-D-galactopyranoside

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17BrO6
CAS Number:
Molecular Weight:
385.21
Beilstein:
92987
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

SMILES string

OC[C@H]1O[C@@H](Oc2ccc3cc(Br)ccc3c2)[C@H](O)[C@@H](O)[C@H]1O

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service