Skip to Content
Merck
All Photos(1)

Documents

807931

Sigma-Aldrich

Copper

nanowires, diam. × L ~100 nm (±20 nm) × 20~30 μm, ~5 mg/mL (in IPA)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cu
Molecular Weight:
63.55
UNSPSC Code:
12352200
NACRES:
NA.23

form

nanowires

concentration

~5 mg/mL (in IPA)

diam. × L

~100 nm (±20 nm) × 20~30 μm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Copper nanowires find applications in flexible electronics, transparent electrodes, photocatalysis, solar cells and as shape memory material [1, 2, 3].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service