Skip to Content
Merck
All Photos(1)

Documents

802824

Sigma-Aldrich

Boron nitride nanotubes

avg. diam. 5 nm , nanotube

Synonym(s):

BNNT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BN
CAS Number:
UNSPSC Code:
12352300

form

nanotubes
powder

Quality Level

composition

H-BN and BNH derivatives, 0-25%
BNNT, >50%
Elemental B, <25%

avg. diam.

5 nm , nanotube

diam.

5 nm ± 2 nm

SMILES string

B=N

InChI

1S/BN/c1-2

InChI key

PZNSFCLAULLKQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Boron Nitride Nanotubes (BNNT) exhibit giant stark effect and electric field tunability of the band gap. BNNTs find applications in photovoltaics, nanoelectronics and as additive in polymeric composites. BNNTs show non-cytotoxicity which makes them promising for biomedical, therapeutic and diagnostics applications as well.

Legal Information

Product of Tekna Advanced Materials, Inc
Tekmat is a trademark of Tekna Advanced Materials, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boron Nitride Nanotubes for Drug Delivery.
Nanjwade BK, et al.
Advances in Materials Science Research, 10, 307-307 (2011)
Polymer composites reinforced by nanotubes as scaffolds for tissue engineering.
Wang W, et al.
International Journal of Polymer Science, 2014 (2014)
Polymer composites of boron nitride nanotubes and nanosheets.
Meng W, et al.
Journal of Material Chemistry C, 2(47), 10049-10061 (2014)
Tuğbagül Çal et al.
Daru : journal of Faculty of Pharmacy, Tehran University of Medical Sciences, 27(1), 203-218 (2019-04-04)
Stem cells provide an opportunity to analyse the effects of xenobiotic on cell viability, differentiation and cell functions. Evaluation of the possible cytotoxic and DNA damaging effects on bone marrow CD34+ stem cells is important for their ability to differentiate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service