Skip to Content
Merck
All Photos(1)

Documents

797588

Sigma-Aldrich

7-Amino-5-thia-2-azaspiro[3.4]octane-2-carboxylic acid-5,5-dioxide 1,1-dimethylethyl ester

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20N2O4S
CAS Number:
Molecular Weight:
276.35
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

storage temp.

2-8°C

SMILES string

NC(CS1(=O)=O)CC21CN(C(OC(C)(C)C)=O)C2

InChI

1S/C11H20N2O4S/c1-10(2,3)17-9(14)13-6-11(7-13)4-8(12)5-18(11,15)16/h8H,4-7,12H2,1-3H3

InChI key

HIXKKZAKBLIEPB-UHFFFAOYSA-N

Application

Spirocyclic building block was first synthesized by Carreira and coworkers which provides a new area of chemical space with straightforward functional handles for further diversification.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Spirocyclic modules containing four-membered rings are currently of growing interest to discovery chemists.

Related Content

The Carreira Research Group is focused on expanding and creating access to uncharted landscape in chemical space. In joint efforts with SpiroChem, Carreira develops innovative spirocyclic building blocks, seeking to make them available to the community at large. Molecules constructed from these building blocks take on unique three-dimensional profiles due to the underlying spirocyclic scaffold, enriched by the presence of diverse combinations of exit vectors as sites for functionalization. Importantly, the spirocyclic building blocks possess physicochemical properties useful in the drug discovery process. Thus, drug leads can be tuned through appending these subunits to the periphery of a given scaffold. Moreover, these compact modules represent a useful collection of unprecedented inputs for fragment-based libraries. In all applications, the inherent novelty of the structure affords researchers new opportunities to run wild in their designs and avenues to chemical space – with their imagination as the sole limitations. We are proud to partner in the efforts to make these building blocks widely available.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service