Skip to Content
Merck
All Photos(1)

Key Documents

79582

Sigma-Aldrich

Phosphorus(V) oxychloride

puriss. p.a., ≥99.0% (AT)

Synonym(s):

Phosphorus(V) oxide chloride, Phosphoryl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
POCl3
CAS Number:
Molecular Weight:
153.33
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

vapor density

5.3 (vs air)

Quality Level

vapor pressure

104 mmHg ( 50 °C)
28 mmHg ( 20 °C)

grade

puriss. p.a.

Assay

≥99.0% (AT)

form

liquid

reaction suitability

reagent type: oxidant

bp

105.8 °C (lit.)

mp

1.25 °C (lit.)

density

1.645 g/mL at 25 °C (lit.)

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

ClP(Cl)(Cl)=O

InChI

1S/Cl3OP/c1-5(2,3)4

InChI key

XHXFXVLFKHQFAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phosphorus(V) oxychloride is a chlorinating agent that reacts with dimethylformamide (DMF) to form Vilsmeier reagent used for Vilsmeier-Haack formylation of aromatic compounds.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nitrogen bridgehead compounds. Part 30. Vilsmeier?Haack formylation of 6, 7, 8, 9-tetrahydro-4 H-pyrido [1, 2-a] pyrimidin-4-ones.
Horvath A
Journal of the Chemical Society. Perkin Transactions 1, 369-377 (1983)
Vilsmeier-Haack formylation of 2-(1H-pyrazol-1-yl) ethanol and its methyl derivatives.
Rstakyan VI, et al.
Russ. J. Gen. Chem., 85(5), 1191-1193 (2015)
Acylation of indoles by Duff reaction and Vilsmeier-Haack formylation and conformation of N-formylindoles.
Chatterjee A and Biswas KM.
The Journal of Organic Chemistry, 38(23), 4002-4004 (1973)
Vilsmeier Reagent: An Efficient Reagent for the Transformation of 2-Aminobenzamides into Quinazolin-4 (3 H)-one Derivatives.
Farzipour S, et al.
Synthetic Communications, 44(4), 481-487 (2014)
Regioselective formylation of pyrazolo [3, 4-b] pyridine and pyrazolo [1, 5-a] pyrimidine systems using Vilsmeier?Haack conditions.
Quiroga J
Tetrahedron Letters, 49(17), 2689-2691 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service