Skip to Content
Merck
All Photos(1)

Key Documents

795550

Sigma-Aldrich

Lead(II) iodide solution

0.55 M in DMF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbI2
Molecular Weight:
461.01
UNSPSC Code:
12352302

form

solution

concentration

0.55 M in DMF

impurities

≤0.2% water

density

1.207 at 25 °C

Application

PbI2 is a well know salt and it finds applications in high energy photon detectors.[1] Recently, lead iodide has received attention due to its application in synthesis of perovskite based dye sensitized solar cells.[2] The solution of PbI2 in DMF is quite commonly used in synthesis of perovskite based Organometallic thin films.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1A - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

137.0 °F

Flash Point(C)

58.33 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luminescence and Photosensitivity of PbI2 Crystals.
Novosad SS, et al.
Inorganic Materials, 38(10), 1058-1062 (2002)
Julian Burschka et al.
Nature, 499(7458), 316-319 (2013-07-12)
Following pioneering work, solution-processable organic-inorganic hybrid perovskites-such as CH3NH3PbX3 (X = Cl, Br, I)-have attracted attention as light-harvesting materials for mesoscopic solar cells. So far, the perovskite pigment has been deposited in a single step onto mesoporous metal oxide films

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Controlled synthesis of metal clusters regulates ligands and atoms, advancing metal nanomaterial synthesis.

Next generation solar cells have the potential to achieve conversion efficiencies beyond the Shockley-Queisser (S-Q) limit while also significantly lowering production costs.

Dr. Perini and Professor Correa-Baena discuss the latest research and effort to obtain higher performance and stability of perovskite materials.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service