Skip to Content
Merck
All Photos(1)

Key Documents

795224

Sigma-Aldrich

trans-1-Butenylboronic acid MIDA ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14BNO4
CAS Number:
Molecular Weight:
211.02
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

SMILES string

CC/C=C/B1OC(CN(C)CC(O1)=O)=O

InChI

1S/C9H14BNO4/c1-3-4-5-10-14-8(12)6-11(2)7-9(13)15-10/h4-5H,3,6-7H2,1-2H3/b5-4+

InChI key

VYHBJTPGBOGZLT-SNAWJCMRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

This MIDA Boronate building block could be utilized in the synthesis of complex polyene-containing molecules.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service