Skip to Content
Merck
All Photos(1)

Key Documents

78110

Sigma-Aldrich

Phe-Gly hydrate

≥98.0% (dried material)

Synonym(s):

L-Phenylalanyl-glycine hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NH2)CONHCH2COOH · aq
CAS Number:
Molecular Weight:
222.24 (anhydrous basis)
Beilstein:
2218143
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0% (dried material)

form

crystals

optical activity

[α]20/D +103±3°, c = 1% in H2O

impurities

≤5% ethanol
≤5% water

mp

~260 °C (dec.)

application(s)

peptide synthesis

SMILES string

O.N[C@@H](Cc1ccccc1)C(=O)NCC(O)=O

InChI

1S/C11H14N2O3.H2O/c12-9(11(16)13-7-10(14)15)6-8-4-2-1-3-5-8;/h1-5,9H,6-7,12H2,(H,13,16)(H,14,15);1H2/t9-;/m0./s1

InChI key

QNLAQUFDXQTNOW-FVGYRXGTSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Inui et al.
Biochimica et biophysica acta, 769(2), 449-454 (1984-01-25)
Carrier-mediated transport of aminocephalosporin antibiotics by renal brush-border membrane vesicles has been studied in relation to the transport systems for dipeptides and amino acids. Dipeptides such as L-carnosine (beta-alanyl-L-histidine) and L-phenylalanylglycine competitively inhibited the uptake of cephalexin, but amino acids
Maria Andreasen et al.
Biochimica et biophysica acta, 1824(2), 274-285 (2011-11-09)
The well-ordered cross β-strand structure found in amyloid aggregates is stabilized by many different side chain interactions, including hydrophobic interactions, electrostatic charge and the intrinsic propensity to form β-sheet structures. In addition to the side chains, backbone interactions are important
A H Dantzig et al.
Biochemical and biophysical research communications, 155(2), 1082-1087 (1988-09-15)
A transport carrier for cephalexin, a cephalosporin antibiotic, was identified in a human intestinal cell line, HT-29. Uptake via the carrier was inhibited by dipeptides, phe-gly, gly-pro, carnosine, and cefaclor, a close drug analog. Uptake was unaffected by the presence
S Borg et al.
Journal of medicinal chemistry, 42(21), 4331-4342 (1999-11-02)
Enantiopure heterocyclic Boc-protected Phe-Gly dipeptidomimetics containing 1,3,4-oxadiazole, 1,2,4-oxadiazole, and 1,2,4-triazole ring systems have been synthesized as building blocks in the synthesis of pseudopeptides. Three derivatives (1-3) have the carboxylic acid function directly bound to the heterocyclic ring, and three derivatives
Jon Våbenø et al.
The Journal of organic chemistry, 67(26), 9186-9191 (2002-12-21)
The readily available N-Boc-protected delta-amino alpha,beta-unsaturated gamma-keto ester 1 was diastereoselectively reduced to the corresponding alcohols 2 and 3, using boron- and aluminum-based reducing reagents. Reduction reactions were successful and resulted in anti/syn ratios of alcohols of >95:5 (80% yield)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service