Skip to Content
Merck
All Photos(1)

Key Documents

775800

Sigma-Aldrich

3-Amino-2-bromo-5-fluoropyridine

97%

Synonym(s):

2-Bromo-5-fluoro-pyridin-3-amine, 2-Bromo-5-fluoro-3-pyridinamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4BrFN2
Molecular Weight:
191.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

114-116 °C

functional group

bromo
fluoro

SMILES string

Nc1cc(F)cnc1Br

InChI

1S/C5H4BrFN2/c6-5-4(8)1-3(7)2-9-5/h1-2H,8H2

InChI key

QUZAKZBKMMUARE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service