Skip to Content
Merck
All Photos(1)

Documents

775355

Sigma-Aldrich

Methanol-17O

80 atom % 17O, 98% (CP)

Synonym(s):

Methyl alcohol-17O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH317OH
Molecular Weight:
33.04
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:

isotopic purity

80 atom % 17O

Assay

98% (CP)

form

liquid

bp

65.4 °C (lit.)

mass shift

M+1

SMILES string

C[17OH]

InChI

1S/CH4O/c1-2/h2H,1H3/i2+1

InChI key

OKKJLVBELUTLKV-VQEHIDDOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service