Skip to Content
Merck
All Photos(1)

Documents

774626

Sigma-Aldrich

Oleamide-d35

98 atom % D, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)6CD2(CD)2(CD2)7COND2
Molecular Weight:
316.69
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98%

form

solid

mass shift

M+35

storage temp.

−20°C

SMILES string

O=C(N([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])/C([2H])=C([2H])\C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])(C([2H])([2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H])[2H]

InChI

1S/C18H35NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H2,19,20)/b10-9-/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D,10D,11D2,12D2,13D2,14D2,15D2,16D2,17D2/hD2

InChI key

FATBGEAMYMYZAF-IZJOVACFSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service