Skip to Content
Merck
All Photos(1)

Key Documents

773409

Sigma-Aldrich

Poly(L-lactide), 2-bromoisobutyryl terminated

average Mn 10,000, PDI ≤1.1

Synonym(s):

10 k PLLA-ATRP, ATRP Macromonomer, ATRP terminated PLLA, PLA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(C3H4O2)nC6H10BrO3
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

optical activity

[α]22/D -145°, c = 0.1% in chloroform

mol wt

average Mn 10,000

mp

160-165 °C

PDI

≤1.1

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Macroinitiator for atom transfer radical polymerization (ATRP) to prepare block copolymers. Poly(L-lactide) is commonly used in drug delivery.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oh; Jung Kwon.
Soft Matter, 7, 5096-5096 (2011)

Articles

Local delivery of bioactive molecules using an implantable device can decrease the amount of drug dose required as well as non-target site toxicities compared to oral or systemic drug administration.

Microparticles in drug delivery: Study on controlling chitosan microparticle size and distribution, exploring encapsulation of BSA and TPP cross-linker.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service