Skip to Content
Merck
All Photos(1)

Documents

772968

Sigma-Aldrich

Diisopropylfluorophosphate-d14

≥98 atom % D, ≥97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D14FO3P
Molecular Weight:
198.23
UNSPSC Code:
12352001
PubChem Substance ID:

isotopic purity

≥98 atom % D

Assay

≥97% (CP)

form

liquid

mass shift

M+14

storage temp.

2-8°C

SMILES string

O=P(OC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])(OC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])F

InChI

1S/C6H14FO3P/c1-5(2)9-11(7,8)10-6(3)4/h5-6H,1-4H3/i1D3,2D3,3D3,4D3,5D,6D

InChI key

MUCZHBLJLSDCSD-ZVEBFXSZSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service