Skip to Content
Merck
All Photos(1)

Key Documents

76854

Sigma-Aldrich

(S)-(−)-5,5′-Dichloro-2,2′-bis(diphenylphosphino)-6,6′-dimethoxy-1,1′-biphenyl

≥97.0% (H-NMR)

Synonym(s):

(−)-(5,5′-Dichloro-6,6′-dimethoxy-1,1′-biphenyl)-2,2′-diyl-bis(diphenylphosphine), (−)-2,2′-Bis(diphenylphosphino)-5,5′-dichloro-6,6′-dimethoxy-1,1′-biphenyl, (S)-(−)-Cl-MeO-BIPHEP

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 1’350.00

CHF 1’350.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
CHF 1’350.00

About This Item

Empirical Formula (Hill Notation):
C38H30Cl2O2P2
CAS Number:
Molecular Weight:
651.50
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

CHF 1’350.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97.0% (H-NMR)

form

crystals

optical activity

[α]20/D −59.0±4°, c = 1% in chloroform

functional group

chloro
phosphine

SMILES string

COc1c(Cl)ccc(P(c2ccccc2)c3ccccc3)c1-c4c(OC)c(Cl)ccc4P(c5ccccc5)c6ccccc6

InChI

1S/C38H30Cl2O2P2/c1-41-37-31(39)23-25-33(43(27-15-7-3-8-16-27)28-17-9-4-10-18-28)35(37)36-34(26-24-32(40)38(36)42-2)44(29-19-11-5-12-20-29)30-21-13-6-14-22-30/h3-26H,1-2H3

InChI key

GCBRCSSVOVNEIX-UHFFFAOYSA-N

Legal Information

produced by Bayer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service