766119
5-Fluoro-2-pyridylzinc bromide solution
0.6 M in THF
About This Item
Recommended Products
form
solution
reaction suitability
core: zinc
reagent type: catalyst
reaction type: Negishi Coupling
concentration
0.6 M in THF
impurities
<5% free chloride
density
1.002 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
Fc1ccc([Zn]Br)nc1
InChI
1S/C5H3FN.BrH.Zn/c6-5-2-1-3-7-4-5;;/h1-2,4H;1H;/q;;+1/p-1
InChI key
GUNYTXBFDVSHAJ-UHFFFAOYSA-M
Application
technical-documents
Legal Information
Rieke is a registered trademark of Rieke Metals, Inc.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-2.2 °F
Flash Point(C)
-19 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service