Skip to Content
Merck
All Photos(1)

Key Documents

762121

Sigma-Aldrich

4-Bromobenzocyclobutene

96%

Synonym(s):

3-Bromobicyclo[4.2.0]octa-1,3,5-triene, 4-Bromo-1,2-dihydrobenzocyclobutene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CHF 104.00

CHF 104.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
CHF 104.00

About This Item

Empirical Formula (Hill Notation):
C8H7Br
CAS Number:
Molecular Weight:
183.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CHF 104.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.589

density

1.470 g/mL at 25 °C

functional group

bromo

storage temp.

2-8°C

SMILES string

Brc1ccc2CCc2c1

InChI

1S/C8H7Br/c9-8-4-3-6-1-2-7(6)5-8/h3-5H,1-2H2

InChI key

GMHHTGYHERDNLO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F

Flash Point(C)

100 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service